Learn More
Sigma Aldrich Fine Chemicals Biosciences Human C Reactive Protein E
Catalog No. 502904873
Encompass
The Human CRP (C Reactive Protein) ELISA (Enzyme- Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CRP in serum plasma cell culture supernatants and urine.
Catalog No. 50-290-4873 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. RAB00961KT
The Human CRP (C Reactive Protein) ELISA (Enzyme- Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human CRP in serum plasma cell culture supernatants and urine.