Learn More
Gibco™ Human C1 Inhibitor Recombinant Protein, PeproTech®
Description
Recombinant Human C1 Inhibitor is a highly glycosylated glycoprotein containing 478 amino acid residues (52.8kDa), corresponding to amino acids 23-500 of the C1 inhibitor precursor, and is fully functional in its ability to inhibit the C1 complex. Glycosylated C1 Inhibitor migrates at an apparent molecular weight of approximately 80-90 kDa by SDS PAGE analysis under reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P05155 |
| For Use With (Application) | Bioactivity, Control |
| Formulation | Protein with no preservative |
| Gene ID (Entrez) | 710 |
| Molecular Weight (g/mol) | 52.8 kDa |
| Name | Human C1 Inhibitor |
| Purification Method | Purified |
| Quantity | 200 μg |
| Source | CHO cells |
| Storage Requirements | -20°C |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.