Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human C19orf52 (aa 164-243) Control Fragment Recombinant Protein

Catalog No. RP99123
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RP99123 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RP99123 Supplier Invitrogen™ Supplier No. RP99123
  • $283.00
    $212.65 / Each of 1

    Save $70.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59759. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

TIMM29, also known as C19orf52, is identified as a novel subunit of the human TIM22 complex, which plays a critical role in the mitochondrial inner membrane. This protein is crucial for the stability of the TIM22 complex, which is responsible for the translocation of hydrophobic inner mitochondrial membrane proteins carrying internal targeting signals. TIMM29/C19orf52 is integrated into the mitochondrial inner membrane, exposing its C-terminus to the intermembrane space, and is vital for mitochondrial function and protein import pathways. This gene's importance is underscored by its involvement in processes related to mitochondrial integrity and its potential role in neurodegenerative disorders and viral interactions, such as those with the hepatitis B virus (HBV).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BSF4
Concentration ≥5.0 mg/mL
For Use With (Application) Neutralization, Control
Formulation PBS, 1M urea with no preservative; pH 7.4
Gene ID (Entrez) 90580
Name Human C19orf52 (aa 164-243) Control Fragment
pH Range 7.4
Purification Method Purified
Quantity 100 μL
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Gene Alias 1810026J23Rik; C19orf52; chromosome 19 open reading frame 52; Mitochondrial import inner membrane translocase subunit Tim29; RGD1309188; RIKEN cDNA 1810026J23 gene; similar to hypothetical protein BC011833; TIM29; TIMM29; uncharacterized protein C19orf52; uncharacterized protein C19orf52 homolog
Common Name C19orf52
Gene Symbol TIMM29
Product Type Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence LDVGFVGRWWVLGAWMRDCDINDDEFLHLPAHLRVVGPQQLHSETNERLFDEKYKPVVLTDDQVDQALWEEQVLQKEKKD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.