Learn More
Biomatik Corporation Human C1INH (Complement 1 Inhibitor) ELISA Kit
Supplier:  Biomatik Corporation EKF58821
Encompass_Preferred
Human C1INH (Complement 1 Inhibitor) ELISA Kit for detecting C1INH in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.125-200ng/ml. Sensitivity: 1.875ng/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: P05155. Target Synonyms: C1INH; SERPING1; C1IN; C1INH; C1NH; HAE1; HAE2; C1 Inhibitor; C1IN; C1INH; C1NH; HAE1; HAE2; SERPING1 serpin peptidase inhibitor; clade G; C1 inhibitor; member 1; Plasma protease C1 inhibitor; Serpin G1; C1 esterase inhibitor; C1-inhibiting factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5233