Learn More
Raybiotech Inc Human C1q R1 (CD93) ELISA Kit
Supplier:  Raybiotech Inc ELHC1QR11
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 24 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Complement Component C1q Receptor (CD93)
Catalog No. 50-194-9039