Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human C1R/Complement Component C1r ELISA Kit

Catalog No. EH56RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH56RBX5 5 x 96 Tests
EH56RBX10 10 x 96 Tests
EH56RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH56RBX5 Supplier Invitrogen™ Supplier No. EH56RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human C1R/Complement Component C1r quantitates human C1R/Complement Component C1r in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human C1R/Complement Component C1r.

C1 component of human complement is a complex of three glycoproteins: C1q, C1r and C1s. C1r has a molecular weight of 83,000 and is present in plasma in an average concentration of 50 μg/mL, either as a complex with C1q and C1s or in the absence of C1q as a tetramer with C1s. The subcomponents of the C1 complex are responsible for the initiation of the classical pathway of complement activation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P00736
Assay Range 0.065-50 ng/mL
Assay Sensitivity 0.065 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI132558; C1r; C1ra; complement C1r; complement C1r subcomponent; Complement C1r subcomponent heavy chain; Complement C1r subcomponent light chain; Complement C1r-A subcomponent; Complement C1r-A subcomponent heavy chain; Complement C1r-A subcomponent light chain; Complement component 1 subcomponent r; Complement component 1 subcomponent r-A; complement component 1, r subcomponent; complement component 1, r subcomponent A; complement component 1, r-A subcomponent; mC1rAAI132558; C1r; C1ra; complement C1r; complement C1r subcomponent; Complement C1r subcomponent heavy chain; Complement C1r subcomponent light chain; Complement C1r-A subcomponent; Complement C1r-A subcomponent heavy chain; Complement C1r-A subcomponent light chain; Complement component 1 subcomponent r; Complement component 1 subcomponent r-A; complement component 1, r subcomponent; complement component 1, r subcomponent A; complement component 1, r-A subcomponent; mC1rA;
Gene ID (Entrez) 715
Gene Symbol C1R
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target C1r
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.