Learn More
Sino Biological Anti-Complement C2 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10154MM06H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Component C2 (rh Complement Component C2; Catalog#10154-H08H; NP_000054.2; Met1-Leu752). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-2369