Learn More
Sino Biological Anti-Complement C2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10154MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement Component C2 (rh Complement Component C2; Catalog#10154-H08H; NP_000054.2; Met1-Leu752). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-9121