Learn More
Abcam Human C3a ELISA kit, 1KIT
Supplier:  Abcam AB2793521KIT
Encompass_Preferred
Human C3a ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human C3a in Human in Plasma, Cell culture supernatant, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2547374