Learn More
BIOHIPPO Human C3a Microsample
SDP
Catalog No. 5000314168
Encompass
Human C3a Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C3a in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 7 81-500 ng/mL with a sensitivity of 2 04 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human C3a Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human C3a Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human C3a biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14168 Supplier BIOHIPPO Supplier No. BHE15212654
Human C3a Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C3a in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 7 81-500 ng/mL with a sensitivity of 2 04 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human C3a Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human C3a Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human C3a biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color