Learn More
Biomatik Corporation Human C4 Binding Protein Beta (C4BPb) CLIA Kit
Supplier:  Biomatik Corporation EKU09845
Encompass_Preferred
Human C4 Binding Protein Beta (C4BPb) CLIA Kit for detecting C4BPb in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 41.2-30,000pg/ml. Sensitivity: 17.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P20851. Gene ID: 725. Target Synonyms: C4BP-B; C4-BP; Complement Component 4 Binding Protein, Beta. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8417