Learn More
BIOHIPPO Human C4BPb Microsample
SDP
Catalog No. 5000314402
Encompass
Human C4BPb Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C4BPb in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 14 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human C4BPb Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human C4BPb Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human C4BPb biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14402 Supplier BIOHIPPO Supplier No. BHE15212117
Human C4BPb Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of C4BPb in Serum plasma and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 14 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human C4BPb Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human C4BPb Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human C4BPb biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color