Learn More
Sino Biological Anti-C5a Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10604MM08100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human C5a (rh C5a; Catalog#10604-HNAE; NP_001726.2; Leu679-Arg751). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-241-1232