Learn More
Sino Biological Human C5 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10604MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Complement C5a (rh Complement C5a; Catalog#10604-HNAE; NP_001726.2; Leu 679-Arg 751). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2778