Learn More
Sino Biological Anti-C5a Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10604RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human C5a (rh C5a; Catalog#10604-HNAE; NP_001726.2; Leu 679-Arg 751). C5a specific IgG was purified by human C5a affinity chromatography.
Catalog No. 50-241-1128