Learn More
Sino Biological C5 Protein, Human, Recombinant (His & FLAG Tag)
Supplier:  Sino Biological 13416H18H1
Encompass_Preferred
A DNA sequence encoding the human C5 (NP_001726.2) (Gln19-Cys1676) was expressed with a polyhistidine tag at the C-terminus and a FLAG tag at the N-terminus.
Catalog No. 50-255-4245