Learn More
Raybiotech Inc Human C5a ELISA Kit
Supplier:  Raybiotech Inc ELHCCC5A1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 31 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human C5a, does not cross-react with rhC3a, rha2-macroglobulin, rmC5a, or rmC5d.
Catalog No. 50-194-9068