Learn More
Sino Biological Human C5a Matched ELISA Antibody Pair Set 15 Plates
Catalog No. 5000785477
Encompass_Preferred
The Human C5a ELISA Pair Set is for the quantitative determination of Human C5a.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human C5a coated on a 96-well plate. Standards and samples are added to the wells, and any Human C5a present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human C5a monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human C5a present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-85477 Supplier Sino Biological Supplier No. SEKA1060415
The Human C5a ELISA Pair Set is for the quantitative determination of Human C5a.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human C5a coated on a 96-well plate. Standards and samples are added to the wells, and any Human C5a present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Human C5a monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human C5a present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.