Learn More
Sino Biological Recombinant Human C5AR1/C5R1/CD88 Protein (VLP, Full-Length, Flag Tag)
Catalog No. 502993019
Encompass_Preferred
A DNA sequence encoding the Human C5AR1 (P21730) (Met1-Val350) was expressed with a flag tag at the N-terminus.
Catalog No. 50-299-3019 Supplier Sino Biological Supplier No. 12154H22H100
A DNA sequence encoding the Human C5AR1 (P21730) (Met1-Val350) was expressed with a flag tag at the N-terminus.