Learn More
Sino Biological Recombinant Human C5AR1/C5R1/CD88 Protein (Nanodisc, thermostabilizing mutations, Flag-Strep & His tag)
Catalog No. 502993020
Encompass_Preferred
A DNA sequence encoding the Human C5AR1 (P21730)(Met1-Lys333) was expressed, with a Flag tag followed by a Strep Tag at the N-terminus, a polyhistidine tag at the C-terminus. Within the expressed regions, there are several proprietary thermostabilizing mutations. This protein was prepared using our Nanodisc platform. Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.
Catalog No. 50-299-3020 Supplier Sino Biological Supplier No. 12154H91HNA20
A DNA sequence encoding the Human C5AR1 (P21730)(Met1-Lys333) was expressed, with a Flag tag followed by a Strep Tag at the N-terminus, a polyhistidine tag at the C-terminus. Within the expressed regions, there are several proprietary thermostabilizing mutations. This protein was prepared using our Nanodisc platform. Nanodisc is a versatile tool for studying membrane proteins. Using styrene-maleic acid (SMA) copolymer, membrane proteins can be extracted directly from prokaryotic and eukaryotic expression systems in the absence of detergents to preserve the protein structure and function better. Compared to membrane scaffold proteins (MSPs) nanodiscs, SMA nanodiscs also have the advantage of preserving proteins' nature by maintaining native lipids surrounded without introducing any heterologous proteins, which allows studies of protein structure and functions in a native-like environment.