Learn More
Sino Biological Human C6 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10587MM05P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human C6 (rh C6; Catalog#12426-H08H; AAA59668.1; Met1-Ala934). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-4400