Learn More
Sino Biological Human C7orf26 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 204640T12100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human C7orf26 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-2945