Learn More
Sino Biological Anti-Carbonic Anhydrase X Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10687MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CARPX (rh CARPX; Catalog#10687-H08H; NP_001076002.1; Met 1-Asn 300). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8089