Learn More
Sino Biological Human CA10 Protein 1MG (Sino Biological)
Supplier:  Sino Biological 10687H08H1
Encompass_Preferred
A DNA sequence encoding the human CARPX (NP_001076002.1) (Met 1-Asn 300) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-237-3770