Learn More
Sino Biological Anti-carbonic anhydrase XII/CA12 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10617MM11P100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Carbonic Anhydrase XII / CA12 (rh Carbonic Anhydrase XII / CA12; Catalog#10617-H08H; NP_001209.1; Met1-Gln291) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-240-9285