Learn More
Sino Biological Recombinant Human carbonic anhydrase XII/CA12 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10617H49HB100
Encompass_Preferred
A DNA sequence encoding the Human CA12 (NP_001209.1)(Met1-Gln291) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-241-0375