Learn More
ACROBiosystems Human CA125 (13810-14451), His Tag (CA5-H52H6) is expressed from human 293 cells (HEK293). It contains AA Pro 13810 - Pro 14451 (Accession # Q8WXI7-1).
Catalog No. 502231402
Encompass_Preferred
The CA125, also known as the MUC16, is a mucin protein that may be found in type I transmembrane or secreted forms that are used monitor the progress of epithelial ovarian cancer therapy. The CA 125 molecule is almost certainly a glycoprotein with a predominance of O-linkages. It is heterogeneous with regard to both size and charge, most likely due to continuous deglycosylation of side chains during its life-span in bodily fluids. It exists as a very large complex (perhaps as much as 4 million daltons) under natural conditions.
This item is not returnable. View return policy
Catalog No. 50-223-1402 Supplier ACROBiosystems Supplier No. CA5H52H61MG
The CA125, also known as the MUC16, is a mucin protein that may be found in type I transmembrane or secreted forms that are used monitor the progress of epithelial ovarian cancer therapy. The CA 125 molecule is almost certainly a glycoprotein with a predominance of O-linkages. It is heterogeneous with regard to both size and charge, most likely due to continuous deglycosylation of side chains during its life-span in bodily fluids. It exists as a very large complex (perhaps as much as 4 million daltons) under natural conditions.