Learn More
Abcam Human CA125 ELISA Kit (MUC16), 10 x 96 Tests
Catalog No. 502655916
Encompass_Preferred
Human CA125 ELISA Kit (MUC16) is a single-wash 90-min Simplestep used to quantify Human CA125 (MUC16) with a sensitivity of 0.02 U/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-5916 Supplier Abcam Supplier No. AB27440210X96TESTS
Add to Cart
Add to Cart
Request a Quote
Human CA125 ELISA Kit (MUC16) is a single-wash 90-min Simplestep used to quantify Human CA125 (MUC16) with a sensitivity of 0.02 U/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement