Learn More
BIOHIPPO Human CA125 Microsample
SDP
Catalog No. 5000315843
Encompass
Human CA125 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CA125 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 U/mL with a sensitivity of 1 11 U/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CA125 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CA125 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CA125 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15843 Supplier BIOHIPPO Supplier No. BHE15212051
Human CA125 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CA125 in Serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 3 13-200 U/mL with a sensitivity of 1 11 U/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CA125 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CA125 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CA125 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color