Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human CA125/MUC16 DuoSet ELISA
SDP

Catalog No. DY560905
Change view
Click to view available options
Quantity:
5 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY560905 5 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY560905 Supplier R&D Systems™ Supplier No. DY560905

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sufficient for at least five 96-well plates

The Human CA125/MUC16 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human CA125/MUC16.

Specifications

Detectable Analytes CA125/MUC16
Assay ELISA Development Kit
Conjugate Biotin
Cross Reactivity < 50% cross-species reactivity observed with species tested. < 0.5% cross-reactivity observed with available related molecules.
Immunoassay Kit Format 96-well microplate (sold separately)
For Use With (Application) ELISA
Gene ID (Entrez) 94025
Host Species Human
Measuring Range 31.2-2,000pg/mL
Quantity 5 Plates
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.