Learn More
Raybiotech Inc Human CA13 ELISA Kit
Supplier:  Raybiotech Inc ELHCA131
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 24 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human CA13. Other species not determined.
Catalog No. 50-194-9049