Learn More
Sino Biological Human CA13 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10461RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase XIII / CA13 (rh Carbonic Anhydrase XIII / CA13; Catalog#10461-H08E; NP_940986.1; Met 1-His 262). Carbonic Anhydrase XIII / CA13 specific IgG was purified by Human Carbonic Anhydrase XIII / CA13 affinity chromatography.
Catalog No. 50-237-3953