Learn More
Sino Biological Anti-Carbonic anhydrase II/CA2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10478T52100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Carbonic anhydrase II (rh Carbonic anhydrase II; Catalog#10478-H08E; NP_000058.1; Met1-Lys260). Carbonic anhydrase II specific IgG was purified by Human Carbonic anhydrase II affinity chromatography.
Catalog No. 50-255-5280