Learn More
Sino Biological Anti-Carbonic anhydrase II/CA2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10478RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase II / CA2 (rh Carbonic Anhydrase II / CA2; Catalog#10478-H08E; NP_000058.1; Met 1-Lys 260). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8621