Learn More
Sino Biological Anti-Carbonic Anhydrase III / CAIII Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10503RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase III (rh Carbonic Anhydrase III; Catalog#10503-H08E; NP_005172.1; Met1-Lys260). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-0868