Learn More
Sino Biological Human CA3 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 10503RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase III / CA3 (rh Carbonic Anhydrase III / CA3; Catalog#10503-H08E; Met 1-Lys 260; NP_005172.1). Carbonic Anhydrase III / CA3 specific IgG was purified by Human Carbonic Anhydrase III / CA3 affinity chromatography.
Catalog No. 50-237-2735