Learn More
Sino Biological Recombinant Human Carbonic Anhydrase IV/CA4 Protein (His Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 10472H08SB20
Encompass_Preferred
A DNA sequence encoding the human CA4 (NP_000708.1) (Met1-Lys283) without the pro peptide was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-241-1605