Learn More
BIOHIPPO Human CA50 Microsample
SDP
Catalog No. 5000314275
Encompass
Human CA50 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CA50 in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 2 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CA50 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CA50 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CA50 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14275 Supplier BIOHIPPO Supplier No. BHE15212189
Human CA50 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of CA50 in Serum plasma and other biological fluids It provides a detection range of 3 13-200 ng/mL with a sensitivity of 1 2 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human CA50 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human CA50 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human CA50 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color