Learn More
Sino Biological Anti-Carbonic Anhydrase 8/CA8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10469RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Carbonic Anhydrase VIII / CA8 (rh Carbonic Anhydrase VIII / CA8; Catalog#10469-H08E; NP_004047.3; Met 1-Gln 290). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-241-2372