Learn More
Sino Biological Recombinant Human Carbonic Anhydrase IX/CA9 Protein (ECD, hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10107H41HB100
Encompass_Preferred
A DNA sequence encoding the Human CA9 (NP_001207.2)(Met1-Asp414) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8450