Learn More
Raybiotech Inc Human Cadherin-13 ELISA Kit
Supplier:  Raybiotech Inc ELHCDH131
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 40 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Cadherin-13 (CDH13)
Catalog No. 50-194-9107