Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Cadherin 13 ELISA Kit

Catalog No. EH60RBX5
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH60RBX5 5 x 96 Tests
EH60RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EH60RBX5 Supplier Invitrogen™ Supplier No. EH60RBX5
Get Up to $3,000 in Extra Life Sciences Consumables with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human Cadherin 13 quantitates human Cadherin 13 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Cadherin 13.

The protein encoded by this gene has a C-terminal domain with tRNA 3 processing endoribonuclease activity, which catalyzes the removal of the 3' trailer from precursor tRNAs. The protein also interacts with activated Smad family member 2 (Smad2) and its nuclear partner forkhead box H1 (also known as FAST-1), and reduced expression can suppress transforming growth factor-beta induced growth arrest. Mutations in this gene result in an increased risk of prostate cancer. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P55290
Assay Range 40-10000 pg/mL
Assay Sensitivity 40 pg/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 4932416G01Rik; CAD13; cadherin 13; cadherin 13, H-cadherin (heart); cadherin-13; CD39 antigen-like 3; CD39L3; CD39-like 3; CDH13; CDHH; Cdht; ecto-apyrase 3; ecto-ATPase 3; ecto-ATPDase 3; HB6; H-cadherin; H-cadherin (heart); heart cadherin; heart-cadherin; NTPDase 3; NTPDase-3; P105; T cad; T Cadherin; Tcad; T-cad; T-cadherin; Truncated cadherin; truncated-cadherin4932416G01Rik; CAD13; cadherin 13; cadherin 13, H-cadherin (heart); cadherin-13; CD39 antigen-like 3; CD39L3; CD39-like 3; CDH13; CDHH; Cdht; ecto-apyrase 3; ecto-ATPase 3; ecto-ATPDase 3; HB6; H-cadherin; H-cadherin (heart); heart cadherin; heart-cadherin; NTPDase 3; NTPDase-3; P105; T cad; T Cadherin; Tcad; T-cad; T-cadherin; Truncated cadherin; truncated-cadherin;
Gene ID (Entrez) 1012
Gene Symbol CDH13
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target H-cadherin
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.