Learn More
ACROBiosystems Human Cadherin-17 1mg
Supplier:  ACROBiosystems CA7H52581MG
Encompass_Preferred
Human Cadherin-17 Protein Fc Tag (CA7-H5258) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Met 787 (Accession # Q12864-1).
Catalog No. 50-253-5441