Learn More
ACROBiosystems Human Cadherin-17 200ug
Catalog No. 502535440
Encompass_Preferred
Biotinylated Human Cadherin-17 Protein FcAvitag (CA7-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Met 787 (Accession # Q12864-1).
Catalog No. 50-253-5440 Supplier ACROBiosystems Supplier No. CA7H82F9200UG
Biotinylated Human Cadherin-17 Protein FcAvitag (CA7-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Met 787 (Accession # Q12864-1).