Learn More
ACROBiosystems Human Cadherin-17 25ug
Supplier:  ACROBiosystems CA7H82F925UG
Encompass_Preferred
Biotinylated Human Cadherin-17 Protein FcAvitag (CA7-H82F9) is expressed from human 293 cells (HEK293). It contains AA Gln 23 - Met 787 (Accession # Q12864-1).
Catalog No. 50-253-5439