Learn More
ACROBiosystems Human Cadherin-3, His Tag (CA3-H52H3) is expressed from human 293 cells (HEK293). It contains AA Asp 108 - Gly 654 (Accession # P22223-1).
Supplier:  ACROBiosystems CA3H52H3100UG
Encompass_Preferred
Cadherin-3 (also known as P-cadherin)is a classical cell-to-cell adhesion molecule with a homeostatic function in several normal tissues. In humans, cadherin-3 is only detected in a few organs. However, its overexpression is strongly associated with a poor prognosis in some solid tumors including breast, lung and pancreatic cancers.
Catalog No. 50-223-0638