Learn More
ACROBiosystems Human Cadherin-6 Protein, His,Avitag™, 25 μg
Catalog No. 502535443
Encompass_Preferred
Cadherin-6 (CDH6), also known as Kidney cadherin or KCAD, is a type II classical cadherin belonging to the cadherin superfamily. These calcium-dependent cell adhesion proteins preferentially interact in a homophilic manner, contributing to the sorting of heterogeneous cell types. This protein is expressed in the human HEK293 system and contains five cadherin domains covering residues Thr 19 to Ala 615. It is supplied as a lyophilized powder with a calculated molecular weight of 70.0 kDa.
  • Greater than 85% purity as determined by SDS-PAGE and 90% by SEC-MALS
  • Features a C-terminal polyhistidine tag followed by an Avi tag
  • Enzymatically biotinylated at the single lysine residue in the Avi tag
  • Expressed from human HEK293 cells
  • Stable for 12 months at -20°C to -70°C in lyophilized state
Catalog No. 50-253-5443 Supplier ACROBiosystems Supplier No. CA6H82E325UG
Cadherin-6 (CDH6), also known as Kidney cadherin or KCAD, is a type II classical cadherin belonging to the cadherin superfamily. These calcium-dependent cell adhesion proteins preferentially interact in a homophilic manner, contributing to the sorting of heterogeneous cell types. This protein is expressed in the human HEK293 system and contains five cadherin domains covering residues Thr 19 to Ala 615. It is supplied as a lyophilized powder with a calculated molecular weight of 70.0 kDa.
  • Greater than 85% purity as determined by SDS-PAGE and 90% by SEC-MALS
  • Features a C-terminal polyhistidine tag followed by an Avi tag
  • Enzymatically biotinylated at the single lysine residue in the Avi tag
  • Expressed from human HEK293 cells
  • Stable for 12 months at -20°C to -70°C in lyophilized state