Learn More
ACROBiosystems Human Cadherin-6 2Vials
Catalog No. 502535269
Encompass_Preferred
Cadherin-6 (CDH6) is also known as Kidney cadherin (K-cadherin or KCAD) is a a type II classical cadherin from the cadherin superfamily. Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. CDH6 / KCAD contains five cadherin domains. CDH6 is highly expressed in brain cerebellum and kidney. Lung pancreas and gastric mucosa show a weak expression and also expressed in certain liver and kidney carcinomas.
Catalog No. 50-253-5269 Supplier ACROBiosystems Supplier No. CHEKATP1272VIALS
Cadherin-6 (CDH6) is also known as Kidney cadherin (K-cadherin or KCAD) is a a type II classical cadherin from the cadherin superfamily. Cadherins are calcium-dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types. CDH6 / KCAD contains five cadherin domains. CDH6 is highly expressed in brain cerebellum and kidney. Lung pancreas and gastric mucosa show a weak expression and also expressed in certain liver and kidney carcinomas.