Learn More
ACROBiosystems ACROBiosystems Human Cadherin-6 / KCAD / CDH6 Protein, Fc Tag (MALS verified)
Catalog No. 502106255
Encompass_Preferred
Recombinant Protein;1MG;Human Cadherin-6 Protein, Fc Tag (CA6-H5258) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Ala 615 (Accession # P55285-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 92.8 kDa. The protein migrates as 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-6255 Supplier ACROBiosystems Supplier No. CA6H52581MG
Recombinant Protein;1MG;Human Cadherin-6 Protein, Fc Tag (CA6-H5258) is expressed from human 293 cells (HEK293). It contains AA Thr 19 - Ala 615 (Accession # P55285-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 92.8 kDa. The protein migrates as 100-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.