Learn More
ACROBiosystems Human Cadherin-9 (CDH9), His-tagged, lyophilized, 64.5 kDa, >90% purity, expressed in HEK293 cells, stored at -20°C, stability 12 months
Catalog No. NC3451159
Encompass_Preferred
Human Cadherin-9 (CDH9), His-tagged, lyophilized, 64.5 kDa, >90% purity, expressed in HEK293 cells, stored at -20°C, stability 12 months
Catalog No. NC3451159 Supplier ACROBiosystems Supplier No. CA9H52H6
Human Cadherin-9 (CDH9), His-tagged, lyophilized, 64.5 kDa, >90% purity, expressed in HEK293 cells, stored at -20°C, stability 12 months